Anti-PCYT2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-11663R-A350
BOSSBS-11663R-A350EA 893 EUR
BOSSBS-11663R-A350
Anti-PCYT2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Phosphatidylethanolamine (PtdEtn) is a major membrane phospholipid which serves to play a primary role in cell membrane structure and is also involved in cell division, cell signaling, activation, phagocytosis and autophagy. PCYT2 (Phosphorylethanolamine transferase), also known as Ethanolamine-phosphate cytidylyltransferase, is a 389 amino acid protein that catalyzes the formation of CDP-ethanolamine from ethanolamine. This product combined with diacylglycerol form phosphatidylethanolamine via the de novo Kennedy pathway. PCYT2 is expressed at highest levels in heart, liver and skeletal muscle. Elevated levels of MyoD, reduced content of Sp1 and a changed ratio of Sp1 to Sp3 all together stimulate upregulation of PCTY2 transcription during C2C12 muscle cell differentiation. Disruption of the PCYT2 gene in mice leads to death after embryo implantation, establishing the necessity of PCYT2 for murine development.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PCYT2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR