Anti-GATB Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Lieferant: Bioss
BS-11667R-A350
BOSSBS-11667R-A350EA 893 EUR
BOSSBS-11667R-A350
Anti-GATB Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
PET112L is the human homolog of the S. cerevisiae COX assembly protein pet112, a protein that is believed to play an important role in the translation of mitochondrial genes. PET112L, also known as HSPC199 or Glu-ADT subunit B (glutamyl-tRNA(Gln) amidotransferase subunit B), is a 557 amino acid protein belonging to the gatB/gatE family of proteins (GatB subfamily) and is believed to play a role in energy metabolism. Localizing to mitochondria, PET112L is expressed in tissues such as heart and muscle, which exhibit high rates of oxidative phosphorylation. The gene encoding PET112L is overexpressed in recurrent ependymoma.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PET112L
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat