Anti-FLAD1 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-11691R
BOSSBS-11691REA 725 EUR
BOSSBS-11691R
Anti-FLAD1 Rabbit Polyclonal Antibody
Antikörper
FLAD1 is a 587 amino acid protein where its N-terminus belongs to the moaB/mog family and its C-terminus belongs to the PAPS reductase family. Existing as five alternatively spliced isoforms, FLAD1 localizes to the cytoplasm and utilizes magnesium as a cofactor. FLAD1 is a key enzyme in the metabolic pathway that converts riboflavin into the redox cofactor flavin adenine dinucleotide (FAD). It is suggested that the molybdenum cofactor biosynthesis protein-like region of FLAD1 may not be functional.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: FLAD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR