Anti-HTT Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-11699R
BOSSBS-11699REA 725 EUR
BOSSBS-11699R
Anti-HTT Rabbit Polyclonal Antibody
Antikörper
May play a role in microtubule-mediated transport or vesicle function.

IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: HTT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR