Anti-AIP1 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-11705R
BOSSBS-11705REA 725 EUR
BOSSBS-11705R
Anti-AIP1 Rabbit Polyclonal Antibody
Antikörper
AIP1 (ASK-interacting protein 1), also known as DAB2IP (DAB2 interacting protein), is a 1,189 amino acid protein that localizes to both the membrane and the cytoplasm and contains one Ras-GAP domain, one PH domain and one C2 domain. Expressed at low levels in prostate tissue, AIP1 functions as a Ras GTPase-activating protein that interacts with ASK 1 and, via this interaction, disrupts the association of ASK 1 with an inhibitory 14-3-3 complex, thereby allowing free ASK 1 to function within the cell. AIP1 exists as multiple alternatively spliced isoforms and is down-regulated in prostate cancer tissue, suggesting a role in tumor suppression. Chromosomal aberrations in the gene encoding AIP1 are associated with acute myeloid leukemia (AML), implicating a role for AIP1 fusion proteins in tumorigenesis.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: AIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR