Anti-CLN8 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-11715R-CY7
BOSSBS-11715R-CY7EA 893 EUR
BOSSBS-11715R-CY7
Anti-CLN8 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
CLN8, a 286 amino acid transmembrane protein, localizes mainly to the endoplasmic reticulum, but also partially to the ER-Golgi intermediate compartment (ERGIC). Mutations in the CLN8 gene cause neuronal ceroid lipofuscinosis 8 and progressive epilepsy with mental retardation (EPMR). Both disorders are forms of neuronal ceroid-lipofuscinose (NCL), a group of progressive neurodegenerative diseases found in children, characterized by failure of psychomotor development, impaired vision, seizures and premature death. The CLN8 protein is one of eight proteins in the CLN family, including CLN1-CLN7, which are associated with NCL.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CLN8
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR