Anti-Calcyon/Drd1ip Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-11719R-A750
BOSSBS-11719R-A750EA 890 EUR
BOSSBS-11719R-A750
Anti-Calcyon/Drd1ip Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Calcyon is a single transmembrane protein that interacts with D1 dopamine receptors. Dopamine is a neurotransmitter that regulates synaptic transmission involved in learning and memory. D1 receptors, the most abundant dopamine receptor in the central nervous system, appear to modulate the activity of D2 dopamine receptors, mediate various behavioural responses, and regulate neuron growth and differentiation. Calcyon is present in neuronal cell bodies and processes of the cortex and hippocampus, and it is especially abundant in pyramidal neurons. Interaction of Calcyon with D1 receptors results in a release of intracellular calcium.

Type: Primary
Antigen: Calcyon/Drd1ip
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 3-65/217
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR