Anti-DGCR6 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-11722R-CY5
BOSSBS-11722R-CY5EA 900 EUR
BOSSBS-11722R-CY5
Anti-DGCR6 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Neural crest cell migration to the third and fourth pharyngeal pouches is a critical step in the structural formation of organs that are affected in DiGeorge syndrome. DGCR6 (DiGeorge syndrome critical region 6) is a nuclear protein that plays a role in neural crest cell migration and is located at the DiGeorge syndrome critical region (DGCR) on chromosome 22. Expressed ubiquitously with highest levels in heart, liver and skeletal muscle, DGCR6 shares high homology with the Drosophila gonadal (gdl) protein and with human Laminin ?1, both of which are involved in early tissue development. The gene encoding DGCR6, along with other DGCR genes, is deleted in DiGeorge syndrome; a developmental disorder characterized by improper facial, cardiac and palate formation. Upregulation of DGCR6 is implicated in lung and colon adenocarcinomas, as well as in Burkitt抯 lymphoma and lymphocytes transformed by EBV. Due to a duplication of the ancestral DGCR6 locus, there are two functional, highly homologous copies of the DGCR6 gene (designated DGCR6 and DGCR6L) on chromosome 22.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DGCR6
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR