Anti-LC3A/B Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-11731R-A750
BOSSBS-11731R-A750EA 899 EUR
BOSSBS-11731R-A750
Anti-LC3A/B Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Ubiquitin-like modifier involved in formation of autophagosomal vacuoles (autophagosomes). Whereas LC3s are involved in elongation of the phagophore membrane, the GABARAP/GATE-16 subfamily is essential for a later stage in autophagosome maturation.

Type: Primary
Antigen: LC3A/B
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 31-121/121
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR