Anti-NMNAT1 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-11738R
BOSSBS-11738REA 725 EUR
BOSSBS-11738R
Anti-NMNAT1 Rabbit Polyclonal Antibody
Antikörper
Nicotinamide adenine dinucleotide (NMNAT) is an essential cofactor involved in fundamental processes in cell metabolism. NMNAT plays a key role in NAD(+) biosynthesis, catalysing the condensation of nicotinamide mononucleotide and ATP, and yielding NAD(+) and pyrophosphate. NMNAT appears to be a substrate of nuclear kinases and contains at least three potential phosphorylation sites. The interaction of NMNAT with nuclear proteins is likely to be modulated by phosphorylation. NMNAT is widely expressed with highest levels in skeletal muscle, heart, liver and kidney.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: NMNAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR