Anti-NMNAT1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-11738R-HRP
BOSSBS-11738R-HRPEA 900 EUR
BOSSBS-11738R-HRP
Anti-NMNAT1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Nicotinamide adenine dinucleotide (NMNAT) is an essential cofactor involved in fundamental processes in cell metabolism. NMNAT plays a key role in NAD(+) biosynthesis, catalysing the condensation of nicotinamide mononucleotide and ATP, and yielding NAD(+) and pyrophosphate. NMNAT appears to be a substrate of nuclear kinases and contains at least three potential phosphorylation sites. The interaction of NMNAT with nuclear proteins is likely to be modulated by phosphorylation. NMNAT is widely expressed with highest levels in skeletal muscle, heart, liver and kidney.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: NMNAT1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR