Anti-PNP Rabbit Polyclonal Antibody
Lieferant: Bioss
BS-11739R
BOSSBS-11739REA 719 EUR
BOSSBS-11739R
Anti-PNP Rabbit Polyclonal Antibody
Antikörper
Purine nucleoside phosphorylase (PNP), also designated inosine phosphorylase, forms a homotrimer. It belongs to the PNP/MTAP phosphorylase family of proteins. Human PNP catalyzes the reversible phosphorolysis of ribonucleosides and 2’-deoxyribonucleosides with specificity for guanine, hypoxanthine, and their analogs. PNP deficiency is a rare autosomal recessive genetic disease associated with a severe defect in T-lymphocyte function and neurologic disorder in children, comprising four percent of combined immunodeficiency cases. Children with PNP deficiency are highly prone to infections, autoimmune disorders, neurological impairment, and cancer.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: PNP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat