Anti-PPT2 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-11750R
BOSSBS-11750REA 725 EUR
BOSSBS-11750R
Anti-PPT2 Rabbit Polyclonal Antibody
Antikörper
PPT2 (palmitoyl-protein thioesterase 2), also known as G14, is a 302 amino acid glycosylated protein that localizes to the lysosome and belongs to the palmitoyl-protein thioesterase family. Expressed throughout the body with highest levels in skeletal muscle, PPT2 functions to remove thioester-linked fatty acyl groups from a variety of substrates, including S-palmitoyl-CoA, thereby playing an important role in lipid metabolism. PPT2 operates at an optimal pH of 7 and exhibits the highest activity for the acyl groups on myristic and palmitic acids, with lower levels of activity toward other short- and long-chain acyl substrates. PPT2 exists as two isoforms, one of which is expressed at low levels and is catalytically inactive.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: PPT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR