Anti-SCGN Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-11754R-CY5
BOSSBS-11754R-CY5EA 900 EUR
BOSSBS-11754R-CY5
Anti-SCGN Rabbit Polyclonal Antibody (Cy5®)
Antikörper
SCGN is a 276 amino acid cytoplasmic protein that contains six EF-hand domains and is related to the calicium-binding proteins Calretinin and Calbindin D28K. Expressed in a variety of tissues including stomach, thyroid, colon, brain and neuroendocrine cells, SCGN is thought to be involved in cell proliferation and KCl (potassium chloride)-mediated calcium flux events. Through its interaction with KCl and its subsequent ability to modulate calcium storage pools within the cell, SCGN may function to negatively control growth and differentiation rates and, thus, indirectly inhibit cell replication.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SCGN
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat