Anti-UBE3A/E6-AP Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-11776R-A680
BOSSBS-11776R-A680EA 890 EUR
BOSSBS-11776R-A680
Anti-UBE3A/E6-AP Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
E6-associating protein is a component of the ubiquitin-mediated proteolytic pathway, which selectively targets proteins for degradation by the 26S proteasome. Ubiquitin (Ub) is directly conjugated to protein substrates by the transfer of Ub from an E2 ubiquitin conjugating enzyme to the target protein. This conjugation is facilitated by the enzymatic activity of E3 ubiquitin ligase family members such as E6-AP. Several substrates of E6-AP have been identified and include the tumour suppressor protein p53 and the mammalian homolog of Rad23, HHR23A. Previous studies have indicated that E6-AP associates with the human papillomavirus E6 oncogene, which complexes with p53 and thereby potentiates E6-AP mediated ubiquitination of p53. Genetic mutations that impair E6-AP activity result in the accumulation of p53 in the cytoplasm, and, in many instances, these mutations are associated with the development of the rare neurodevelopmental disorder Angelman syndrome (AS), which is characterised by severe motor dysfunction and mental retardation.

Type: Primary
Antigen: UBE3A/E6-AP
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 701-800/875
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR