Anti-ALDH3A2 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-11797R-CY5
BOSSBS-11797R-CY5EA 900 EUR
BOSSBS-11797R-CY5
Anti-ALDH3A2 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Aldehyde dehydrogenases (ALDHs) mediate the NADP+-dependent oxidation of aldehydes into acids and play an important role in the detoxification of alcohol-derived acetaldehyde, as well as in lipid peroxidation and in the metabolism of corticosteroids, biogenic amines and neurotransmitters. ALDH3A2 (aldehyde dehydrogenase 3 family, member A2), also known as SLS, FALDH or ALDH10, is a 485 amino acid single-pass membrane protein that localizes to the cytoplasmic side of the endoplasmic reticulum and belongs to the aldehyde dehydrogenase family. Expressed in a variety of tissues, including liver, heart, lung, brain, kidney and placenta, ALDH3A2 catalyzes the NAD+-dependent oxidation of long-chain aliphatic aldehydes to fatty acids, a process that is necessary for detoxification and lipid metabolism. Defects in the gene encoding ALDH3A2 are the cause of Sjoegren-Larsson syndrome (SLS), an autosomal recessive neurocutaneous disorder characterized by severe mental retardation, seizures and speech defects. Multiple isoforms of ALDH3A2 exist due to alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ALDH3A2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR