Anti-SLCO3A1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-11802R-A555
BOSSBS-11802R-A555EA 893 EUR
BOSSBS-11802R-A555
Anti-SLCO3A1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
The organic anion transporter family of proteins mediate hepatic uptake of cardiac glycosides. OATP-D is a 710 amino acid member of the organic anion transporter protein family. As a multi-pass membrane protein, OATP-D mediates the Na+-independent transport of vasopressin, prostaglandins (PG) E1 and E2, thyroxine (T4), deltorphin II and other organic anions, but not estrone-3-sulfate, DPDPE, taurocholate, DHEAS or digoxin. OATP-D is ubiquitously expressed with highest levels present in leukocytes and spleen. OATP-D is expressed as four isoforms produced by alternative splicing events.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLCO3A1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR