Anti-Adracalin Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)

Lieferant: Bioss
BS-11807R-A750
BOSSBS-11807R-A750EA 890 EUR
BOSSBS-11807R-A750
Anti-Adracalin Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)
Antikörper
Aladin (Adracalin) belongs to a family of WD repeat-containing proteins. These proteins have a wide variety of functions, including signal transduction regulation, RNA processing and transcription. Aladin plays a role in peripheral and central nervous system development. It is widely expressed, with the highest expression seen in pituitary gland, corpus callosum, cerebellum, adrenal gland and gastrointestinal structures. Defects in Aladin cause the autosomal recessive disorder achalasia-addisonianism-alacrima (triple A) syndrome. Triple A syndrome is characterised by achalasia, alacrima and adrenocortico-tropin-resistant adrenal insufficiency. Robust expression in neural systems associated with cognitive, motor and sensory functions is consistent with the myriad of symptoms experienced by patients with triple A syndrome.

Type: Primary
Antigen: Adracalin
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-150/546
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR