Anti-BTD Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-11813R-A488
BOSSBS-11813R-A488EA 893 EUR
BOSSBS-11813R-A488
Anti-BTD Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Biotin, also known as vitamin B7, is an essential water-soluble vitamin that is a cofactor in glucogenesis and in the metabolism of fatty acids and leucine. Biotinidase is a 523 amino acid enzyme that catalyzes the hydrolysis of biocytin to biotin and lysine. Secreted into extracellular space, biotinidase is expressed in liver, heart, placenta, brain, skeletal muscle, pancreas and kidney. Biotinidase contains one carbon-nitrogen hydrolase domain, which is involved in the reduction of organic nitrogen compounds and ammonia production. Defects in the gene encoding biotinidase are the cause of biotinidase deficiency, which is characterized by skin rash, ataxia, seizures, hearing loss, hypotonia and optic atrophy. These symptoms are due to the individual’s inability to reutilize biotin and can, therefore, typically be treated with the addition of free biotin.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BTD
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR