Anti-CSAD Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-11822R
BOSSBS-11822REA 725 EUR
BOSSBS-11822R
Anti-CSAD Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CSAD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR