Anti-DRGX Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Lieferant: Bioss
BS-11827R-A488
BOSSBS-11827R-A488EA 893 EUR
BOSSBS-11827R-A488
Anti-DRGX Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
DRGX is a transcription factor required for the formation of correct projections from nociceptive sensory neurons to the dorsal horn of the spinal cord and normal perception of pain.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DRGX
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat