Anti-DRGX Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-11827R-CY3
BOSSBS-11827R-CY3EA 900 EUR
BOSSBS-11827R-CY3
Anti-DRGX Rabbit Polyclonal Antibody (Cy3®)
Antikörper
DRGX is a transcription factor required for the formation of correct projections from nociceptive sensory neurons to the dorsal horn of the spinal cord and normal perception of pain.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DRGX
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR