Anti-DAGLA Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-11833R-A555
BOSSBS-11833R-A555EA 1350 EUR
BOSSBS-11833R-A555
Anti-DAGLA Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Catalyzes the hydrolysis of diacylglycerol (DAG) to 2-arachidonoyl-glycerol (2-AG), the most abundant endocannabinoid in tissues. Required for axonal growth during development and for retrograde synaptic signaling at mature synapses.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: DAGLA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR