Anti-FEZ2 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-11840R-CY3
BOSSBS-11840R-CY3EA 900 EUR
BOSSBS-11840R-CY3
Anti-FEZ2 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Zygin 2 is a 353 amino acid cytoplasmic protein that belongs to the zygin family. Zygin 2 is found as a difulfide-linked homodimer but may for heterodimers with FEZ1. the plasma membrane to the cytoplasm by activation of PKC zeta. PKC zeta also enhances zygin 1 interaction with UFD2 and polyubiquitination. Expressed mainly in brain, zygin 1 is a mammalian ortholog of Caenorhabditis elegans UNC-76 and is able to restore partial locomotion and axonal fasciculation to C. elegans UNC-76 mutants, which suggest a functional role in axonal outgrowth. As component of the network of molecules thatMore Product Information
Bestellen

TECHNISCHE DATEN

Antigen-Symbol FEZ2
Environmentally Preferable
Antigen-Name Fasciculation and Elongation Protein Zeta 2
Antigen-Synonyme Fasciculation and elongation protein zeta-2|HUM3CL|Zygin II|FEZ2
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Cy3®
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Isotyp IgG
ImmunoChemie Yes
Form liquid
Reinigung Purified by Protein A
Konzentration 1 μg/μl
Kreuzadsorption No
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagertemperatur Store at 4°C for 12 months
Transporttemperatur 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR