Anti-FEZ2 Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-11840R-CY5
BOSSBS-11840R-CY5EA 900 EUR
BOSSBS-11840R-CY5
Anti-FEZ2 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Zygin 2 is a 353 amino acid cytoplasmic protein that belongs to the zygin family. Zygin 2 is found as a difulfide-linked homodimer but may for heterodimers with FEZ1. the plasma membrane to the cytoplasm by activation of PKC zeta. PKC zeta also enhances zygin 1 interaction with UFD2 and polyubiquitination. Expressed mainly in brain, zygin 1 is a mammalian ortholog of Caenorhabditis elegans UNC-76 and is able to restore partial locomotion and axonal fasciculation to C. elegans UNC-76 mutants, which suggest a functional role in axonal outgrowth. As component of the network of molecules thatMore Product Information