Anti-GPM6B Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-11847R-CY7
BOSSBS-11847R-CY7EA 900 EUR
BOSSBS-11847R-CY7
Anti-GPM6B Rabbit Polyclonal Antibody (Cy7®)
Antikörper
GPM6B belongs to the myelin proteolipid protein family and is implicated in neural development. It is expressed in neurons and glia; cerebellar Bergmann glia, in glia within white matter tracts of the cerebellum and cerebrum, and in embryonic dorsal root ganglia.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GPM6B
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR