Anti-GPM6B Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Lieferant: Bioss
BS-11847R-FITC
BOSSBS-11847R-FITCEA 900 EUR
BOSSBS-11847R-FITC
Anti-GPM6B Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
GPM6B belongs to the myelin proteolipid protein family and is implicated in neural development. It is expressed in neurons and glia; cerebellar Bergmann glia, in glia within white matter tracts of the cerebellum and cerebrum, and in embryonic dorsal root ganglia.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GPM6B
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat