Anti-HELT Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-11851R
BOSSBS-11851REA 725 EUR
BOSSBS-11851R
Anti-HELT Rabbit Polyclonal Antibody
Antikörper
The Drosophila hairy and Enhancer of split genes encode basic helix-loop-helix (bHLH) transcriptional repressors that function in the Notch signaling pathway and control segmentation and neural development during embryogenesis. The Notch signaling pathway is thought to maintain stem cells through transcriptional activation of HES/HEY family members to repress tissue-specific transcription factors. HESL (HES-like), also known as Mgn or HELT (HES/HEY-like transcription factor), is a 327 amino acid nuclear protein belonging to the HEY family. Containing a basic helix-loop-helix (bHLH) domain and an Orange domain, HESL self-associates and interacts with HES5 and HRT2. HESL is considered a transcriptional repressor that binds to the canonical E box sequence 5'-CACGCG-3'. HESL exists as two isoforms produced by alternative splicing events.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: HELT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR