Anti-ASH1L Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-11866R-A350
BOSSBS-11866R-A350EA 893 EUR
BOSSBS-11866R-A350
Anti-ASH1L Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Histone methyltransferase specifically methylating 'Lys-36' of histone H3 (H3K36me).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ASH1L
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR