Anti-FRA2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-1188R-A750
BOSSBS-1188R-A750EA 890 EUR
BOSSBS-1188R-A750
Anti-FRA2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Controls osteoclast survival and size. As a dimer with JUN, activates LIF transcription. Activates CEBPB transcription in PGE2-activated osteoblasts.

Type: Primary
Antigen: FRA2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 231-326/326
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR