Anti-NOR1 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-11903R
BOSSBS-11903REA 725 EUR
BOSSBS-11903R
Anti-NOR1 Rabbit Polyclonal Antibody
Antikörper
Nur77 (also designated NGFI-B), Nurr1 (Nur-related factor 1), and NOR-1 (neuron-derived orphan receptor 1) constitute the NGFI-B subfamily within the nuclear receptor superfamily. Ligands for these protein have not been identified, and, therefore, they are designated “orphan nuclear receptors”. Genes of the NGFI-B subfamily are classified as immediate-early genes, which are induced rapidly, but transiently, in response to a variety of stimuli. They have been implicated in cell proliferation, differentiation, and apoptosis. The human NOR-1 gene maps to chromosome 9q, and encodes a protein which is expressed in heart, skeletal muscle, thymus, and spleen as well as in brain, where it is developmentally regulated. There-fore, NOR-1 may be involved in regulating neural differentiation. The NOR-1 gene also undergoes chromosomal translocation with the EWS gene to produce a protein thought to affect pre-mRNA splicing.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: NOR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR