Anti-IL-2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-1191R-A750
BOSSBS-1191R-A750EA 890 EUR
BOSSBS-1191R-A750
Anti-IL-2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Produced by T-cells in response to antigenic or mitogenic stimulation, this protein is required for T-cell proliferation and other activities crucial to regulation of the immune response. Can stimulate B-cells, monocytes, lymphokine-activated killer cells, natural killer cells, and glioma cells.
Type: Primary
Antigen: IL-2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 65-155/155
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat