Anti-OTP Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-11920R
BOSSBS-11920REA 1090 EUR
BOSSBS-11920R
Anti-OTP Rabbit Polyclonal Antibody
Antikörper
Homeodomain (HD) genes are helix-turn-helix transcription factors that play key roles in the specification of cell fates. OTP (orthopedia homeobox) is a 325 amino acid nuclear protein belonging to the paired homeobox family and Bicoid subfamily. OTP is expressed in neurons, which give rise to the paraventricular (PVN), supraoptic (SON), anterior periventricular (aPV) and arcuate (ARN) nuclei. Containing a homeobox DNA-binding domain and a OAR domain, OTP is suggested to be involved in the differentiation of hypothalamic neuroendocrine cells. At early embryonic stages in mice, the expression of SIM2 is downregulated in the absence of OTP, indicating a potential role for OTP as an upstream regulator of SIM2. OTP is highly conserved in evolution and is encoded by a gene located on human chromosome 5, which contains 181 million base pairs and comprises nearly 6% of the human genome.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: OTP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR