Anti-OTP Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Lieferant: Bioss
BS-11920R-A555
BOSSBS-11920R-A555EA 1350 EUR
BOSSBS-11920R-A555
Anti-OTP Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Homeodomain (HD) genes are helix-turn-helix transcription factors that play key roles in the specification of cell fates. OTP (orthopedia homeobox) is a 325 amino acid nuclear protein belonging to the paired homeobox family and Bicoid subfamily. OTP is expressed in neurons, which give rise to the paraventricular (PVN), supraoptic (SON), anterior periventricular (aPV) and arcuate (ARN) nuclei. Containing a homeobox DNA-binding domain and a OAR domain, OTP is suggested to be involved in the differentiation of hypothalamic neuroendocrine cells. At early embryonic stages in mice, the expressionMore Product Information