Anti-GATAD1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-11921R-A647
BOSSBS-11921R-A647EA 876 EUR
BOSSBS-11921R-A647
Anti-GATAD1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Component of some chromatin complex recruited to chromatin sites methylated 'Lys-4' of histone H3 (H3K4me).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GATAD1/ODAG
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR