Anti-RDH10 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-11941R-A555
BOSSBS-11941R-A555EA 893 EUR
BOSSBS-11941R-A555
Anti-RDH10 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Retinol dehydrogenase with a clear preference for NADP. Converts all-trans-retinol to all-trans-retinal. Has no detectable activity towards 11-cis-retinol, 9-cis-retinol and 13-cis-retinol.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RDH10
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR