Anti-RDH10 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Lieferant: Bioss
BS-11941R-A647
BOSSBS-11941R-A647EA 893 EUR
BOSSBS-11941R-A647
Anti-RDH10 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Retinol dehydrogenase with a clear preference for NADP. Converts all-trans-retinol to all-trans-retinal. Has no detectable activity towards 11-cis-retinol, 9-cis-retinol and 13-cis-retinol.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RDH10
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat