Anti-SLC12A6 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-11952R-A488
BOSSBS-11952R-A488EA 893 EUR
BOSSBS-11952R-A488
Anti-SLC12A6 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
The four isoforms of potassium/chloride co-transport channels (KCC) belong to a superfamily of cation-chloride co-transporters involved in cell volume maintenance. Nitric oxide (NO) donors activate KCCs, while inhibitors of the cGMP pathway prevent NO donor activation. The ubiquitously expressed KCC1 contains 12 transmembrane domains with both cytoplasmic N and C terminal domains. KCC2 expression is limited to neuronal tissues by a restrictive element similar to the neuronal-restrictive silencing factor. In neurons, KCC2 expression is correlated with an inhibitory response to GABA, while the absence of KCC2 is necessary for an unusual excitatory response to GABA. Alterations of KCC2 expression in the inferior colliculus of rat brain may be related to seizure susceptibility. Conversely, KCC3 is not suspected to play a major role in epilepsy. The two splice variants of KCC3, KCC3a and KCC3b, are predominantly expressed in brain and kidney, respectively, while KCC4 is expressed in muscle, brain, lung, heart and kidney.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC12A6
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR