Anti-SLC25A12 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-11953R-CY3
BOSSBS-11953R-CY3EA 900 EUR
BOSSBS-11953R-CY3
Anti-SLC25A12 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Calcium signaling in mitochondria is important in order for it to function in response to a variety of extracellular stimuli. Signaling begins with Ca(2+) entry in mitochondria via the Ca(2+) uniporter followed by Ca(2+) activation of three dehydrogenases in the mitochondrial matrix. ARALAR, the neuronal Ca(2+)-binding mitochondrial aspartate-glutamate carrier, has Ca(2+) binding domains facing the extramitochondrial space and functions in the malate-aspartate NADH shuttle (MAS). ARALAR is encoded by the SLC25A12 gene and is expressed in brain and skeletal muscle. ARALAR is required for the synthesis of brain aspartate and N-acetylaspartatemay and plays a role in myelin formation. It is also essential for the transmission of small Ca(2+) signals to mitochondria via an increase in mitochondrial NADH. In addition, ARALAR is implicated in conferring susceptibility to schizophrenia.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC25A12
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR