Anti-RAMP3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Lieferant: Bioss
BS-11972R-A350
BOSSBS-11972R-A350EA 876 EUR
BOSSBS-11972R-A350
Anti-RAMP3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Plays a role in cardioprotection by reducing cardiac hypertrophy and perivascular fibrosis in a GPER1-dependent manner. Transports the calcitonin gene-related peptide type 1 receptor (CALCRL) and GPER1 to the plasma membrane. Acts as a receptor for adrenomedullin (AM) together with CALCRL.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RAMP3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat