Anti-RAMP3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss
BS-11972R-FITC
BOSSBS-11972R-FITCEA 883 EUR
BOSSBS-11972R-FITC
Anti-RAMP3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Plays a role in cardioprotection by reducing cardiac hypertrophy and perivascular fibrosis in a GPER1-dependent manner. Transports the calcitonin gene-related peptide type 1 receptor (CALCRL) and GPER1 to the plasma membrane. Acts as a receptor for adrenomedullin (AM) together with CALCRL.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RAMP3
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR