Anti-Serine/threonine-protein kinase MAK-V Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-11990R-A750
BOSSBS-11990R-A750EA 890 EUR
BOSSBS-11990R-A750
Anti-Serine/threonine-protein kinase MAK-V Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
The HUNK (hormonally upregulated Neu-associated kinase) protein, also designated MAK-V in mouse, has been identified as a novel SNF1-related serine/threonine kinase. The human HUNK gene localizes to chromosome 21q22 and encodes a protein with nucleocytoplasmic distribution and localizes to the centrosome. Overexpression of the HUNK protein associates with approximately 50% of breast carcinomas, and may provide diagnostic-prognostic value as a molecular marker. Serine/threonine-protein kinase SNF1-like kinase 2 (SIK) phosphorylates Ser-794 of IRS1 in insulin-stimulated adipocytes, which may modulate the efficiency of insulin signal transduction. SIK is activated by phosphorylation on Thr-175 by STK11 in complex with STE20-related adapter- and CAB39.

Type: Primary
Antigen: Serine/threonine-protein kinase MAK-V
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 71-115/714
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR