Anti-DDAH1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Lieferant: Bioss
BS-11997R-A350
BOSSBS-11997R-A350EA 893 EUR
BOSSBS-11997R-A350
Anti-DDAH1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
DDAH, a dimethylarginine dimethylaminohydrolase, hydrolyzes dimethyl arginine (ADMA) and monomethyl arginine (MMA), both inhibitors of nitric oxide synthases, and may be involved in in-vivo modulation of nitric oxide production (1,2). Impairment of DDAH causes ADMA accumulation and a reduction in cGMP generation (3). DDAH II, the predominant DDAH isoform in endothelial cells, facilitates the induction of nitric oxide synthesis by all-trans-Retinoic acid (atRA) (4). DDAH proteins are highly expressed in colon, kidney, stomach and liver tissues (1).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DDAH1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat