Anti-DDAH1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-11997R-A680
BOSSBS-11997R-A680EA 890 EUR
BOSSBS-11997R-A680
Anti-DDAH1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
DDAH, a dimethylarginine dimethylaminohydrolase, hydrolyses dimethyl arginine (ADMA) and monomethyl arginine (MMA), both inhibitors of nitric oxide synthases, and may be involved in in-vivo modulation of nitric oxide production. Impairment of DDAH causes ADMA accumulation and a reduction in cGMP generation. DDAH II, the predominant DDAH isoform in endothelial cells, facilitates the induction of nitric oxide synthesis by all-trans-Retinoic acid (atRA) (4). DDAH proteins are highly expressed in colon, kidney, stomach and liver tissues.
Type: Primary
Antigen: DDAH1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 201-285/285
Host: Rabbit
Isotype: IgG
Reactivity: Mouse