Anti-DDAH1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-11997R-CY3
BOSSBS-11997R-CY3EA 900 EUR
BOSSBS-11997R-CY3
Anti-DDAH1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
DDAH, a dimethylarginine dimethylaminohydrolase, hydrolyzes dimethyl arginine (ADMA) and monomethyl arginine (MMA), both inhibitors of nitric oxide synthases, and may be involved in in-vivo modulation of nitric oxide production (1,2). Impairment of DDAH causes ADMA accumulation and a reduction in cGMP generation (3). DDAH II, the predominant DDAH isoform in endothelial cells, facilitates the induction of nitric oxide synthesis by all-trans-Retinoic acid (atRA) (4). DDAH proteins are highly expressed in colon, kidney, stomach and liver tissues (1).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DDAH1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR