Anti-CHRM4/mAChR M4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-11998R-A750
BOSSBS-11998R-A750EA 890 EUR
BOSSBS-11998R-A750
Anti-CHRM4/mAChR M4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
The muscarinic acetylcholine receptor mediates various cellular responses, including inhibition of adenylate cyclase, breakdown of phosphoinositides and modulation of potassium channels through the action of G proteins. Primary transducing effect is inhibition of adenylate cyclase.

Type: Primary
Antigen: CHRM4/mAChR M4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-31/479
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR