Neurotensin (NT) initiates an intracellular response by interacting with the G protein-coupled receptors NTR1 (NTS1 receptor, high affinity NTR) and NTR2 (NTS2 receptor, levocabastine-sensitive neurotensin receptor), and the type I receptor NTR3 (NTS3 receptor, sortilin-1, Gp95). NT has a wide distribution in regions of the brain and in peripheral tissues where NT receptors can contribute to hypotension, hyperglycemia, hypothermia, antinociception and regulation of intestinal motility and secretion. HL-60 cells express NTR1, which can couple to Gq, Gi/o, or Gs. Alternative splicing of rat NTR2 can generate a 5-transmembrane domain variant isoform that is co-expressed with the full-length NTR2 throughout the brain and spinal cord. NTR3 activation in the murine microglial cell line N11 induces MIP-2, MCP-1, IL-1beta and TNF?in an ERK1/2 and Akt kinase-dependent manner.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: Neurotensin Receptor 2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat