Anti-GluA4/Ionotropic Glutamate receptor 4 Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)

Lieferant: Bioss
BS-12009R-A750
BOSSBS-12009R-A750EA 890 EUR
BOSSBS-12009R-A750
Anti-GluA4/Ionotropic Glutamate receptor 4 Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)
Antikörper
Glutamate receptors mediate most excitatory neurotransmission in the brain and play an important role in neural plasticity, neural development and neurodegeneration. Ionotropic glutamate receptors are categorised into NMDA receptors and kainate/AMPA receptors, both of which contain glutamate-gated, cation-specific ion channels. Kainate/AMPA receptors are co-localised with NMDA receptors in many synapses and consist of seven structurally related subunits designated GluR-1 to -7. The kainate/AMPA receptors are primarily responsible for the fast excitatory neuro-transmission by glutamate, whereas the NMDA receptors are functionally characterised by a slow kinetic and a high permeability for Ca2+ ions. The NMDA receptors consist of five subunits: epsilion 1, 2, 3, 4 and one seta subunit. The seta subunit is expressed throughout the brainstem, whereas the four epsilon subunits display limited distribution.

Type: Primary
Antigen: GluA4/Ionotropic Glutamate receptor 4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 351-450/902
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR