Anti-GPR126 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-12027R
BOSSBS-12027REA 725 EUR
BOSSBS-12027R
Anti-GPR126 Rabbit Polyclonal Antibody
Antikörper
Orphan receptor. May be required for normal differentiation of promyelinating Schwann cells and for normal myelination of axons. Signals probably through G-proteins to transiently elevate cAMP levels (By similarity).

IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: GPR126
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR