Anti-LPAR4 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-12074R
BOSSBS-12074REA 725 EUR
BOSSBS-12074R
Anti-LPAR4 Rabbit Polyclonal Antibody
Antikörper
Receptor for lysophosphatidic acid (LPA), a mediator of diverse cellular activities. Transduces a signal by increasing the intracellular calcium ions and by stimulating adenylyl cyclase activity. The rank order of potency for agonists of this receptor is 1-oleoyl- >1-stearoyl- >1-palmitoyl- >1-myristoyl- >1-alkyl- >1-alkenyl-LPA (By similarity).

Recommended Dilutions: Western Blot: 1:100 to 1000; Flow Cytometry: 1:20 to 100; IHC-P: 1:100 to 500; IF(IHC-P): 1:50 to 200.

Type: Primary
Antigen: LPAR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR